Product Details

SNP ID
rs146516387
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:39626909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGGGCTGGGCACTCTTCCCAG[C/G]ATGGCGAGCTGGTTGCTCAGGGCTG
Phenotype
MIM: 609034
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HEYL PubMed Links

Gene Details

Gene
HEYL
Gene Name
hes related family bHLH transcription factor with YRPW motif-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014571.3 636 Silent Mutation ATC,ATG I195M NP_055386.1
XM_005270745.2 636 Missense Mutation ATC,ATG I167M XP_005270802.1

View Full Product Details