Product Details

SNP ID
rs146681870
Assay Type
Functionally Tested
NCBI dbSNP Submissions
7
Location
Chr.1:205381744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCACAGCAAGCTTCAAGCCCA[C/T]TGGGAAACCTTCTTCTCTCCAGCTC
Phenotype
MIM: 610480
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LEMD1 PubMed Links

Gene Details

Gene
LEMD1
Gene Name
LEM domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001552.4 750 Silent Mutation CAA,CAG Q106Q NP_001001552.3
NM_001199050.1 750 Missense Mutation ATG,GTG M154V NP_001185979.1
NM_001199051.1 750 Missense Mutation ATG,GTG M113V NP_001185980.1
NM_001199052.1 750 Silent Mutation CAA,CAG Q65Q NP_001185981.1
XM_011510160.2 750 Missense Mutation ATG,GTG M182V XP_011508462.1
XM_011510161.2 750 Missense Mutation ATG,GTG M182V XP_011508463.1
XM_011510162.2 750 Missense Mutation ATG,GTG M176V XP_011508464.1
XM_011510163.2 750 Missense Mutation ATG,GTG M141V XP_011508465.1
XM_011510164.2 750 UTR 3 XP_011508466.1
XM_011510165.2 750 UTR 3 XP_011508467.1
Gene
LEMD1-AS1
Gene Name
LEMD1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details