Product Details

SNP ID
rs146718362
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:1255450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGGGAGAGTCTGGGGTCTGCTA[C/T]TGAGTTCGTCTTGTGCTTTCTGTTT
Phenotype
MIM: 616593
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM132A PubMed Links

Gene Details

Gene
FAM132A
Gene Name
family with sequence similarity 132 member A
There are no transcripts associated with this gene.

Gene
UBE2J2
Gene Name
ubiquitin conjugating enzyme E2 J2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058167.2 595 Missense Mutation NP_477515.2
NM_194315.1 595 Missense Mutation NP_919296.1
NM_194457.1 595 Missense Mutation NP_919439.1
NM_194458.1 595 Missense Mutation NP_919440.1
XM_005244718.3 595 Missense Mutation XP_005244775.1
XM_005244719.4 595 Missense Mutation XP_005244776.1
XM_006710333.3 595 Missense Mutation XP_006710396.1
XM_011540613.2 595 Missense Mutation XP_011538915.1
XM_011540614.2 595 Missense Mutation XP_011538916.1
XM_011540616.2 595 Missense Mutation XP_011538918.1
XM_011540617.2 595 Missense Mutation XP_011538919.1
XM_017000238.1 595 Missense Mutation XP_016855727.1
XM_017000239.1 595 Missense Mutation XP_016855728.1

View Full Product Details