Product Details

SNP ID
rs146772607
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:35714993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCAGCCACAACCTTCAGATAAAT[A/G]ACAGCATGCTAGCCCTCTGTCTCCA
Phenotype
MIM: 605434
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf216 PubMed Links

Gene Details

Gene
C1orf216
Gene Name
chromosome 1 open reading frame 216
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152374.1 1817 UTR 3 NP_689587.1
Gene
CLSPN
Gene Name
claspin
There are no transcripts associated with this gene.

View Full Product Details