Product Details

SNP ID
rs146924212
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:203771414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCGTCATGCCCTTGCTGACTTTG[A/C]CACAAACCAGACAAAGAGCCAAAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LAX1 PubMed Links

Gene Details

Gene
LAX1
Gene Name
lymphocyte transmembrane adaptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136190.1 548 Missense Mutation ACA,CCA T67P NP_001129662.1
NM_001282878.1 548 Missense Mutation ACA,CCA T7P NP_001269807.1
NM_017773.3 548 Missense Mutation ACA,CCA T83P NP_060243.2
XM_006711397.3 548 Missense Mutation ACA,CCA T83P XP_006711460.1

View Full Product Details