Product Details

SNP ID
rs147009675
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.1:965056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCAACTCCATCGAGAAGTACAA[C/T]CCGAGGACCAACAAGTGGGTGGCCG
Phenotype
MIM: 610770
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KLHL17 PubMed Links
Additional Information
For this assay, SNP(s) [rs115741058] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KLHL17
Gene Name
kelch like family member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198317.2 2454 Silent Mutation AAC,AAT N598N NP_938073.1
XM_006710600.3 2454 Silent Mutation AAC,AAT N621N XP_006710663.1
XM_006710601.3 2454 Intron XP_006710664.1
Gene
NOC2L
Gene Name
NOC2 like nucleolar associated transcriptional repressor
There are no transcripts associated with this gene.

Gene
PLEKHN1
Gene Name
pleckstrin homology domain containing N1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160184.1 2454 Intron NP_001153656.1
NM_032129.2 2454 Intron NP_115505.2
XM_006710944.3 2454 Intron XP_006711007.2
XM_011542248.2 2454 Intron XP_011540550.2
XM_017002474.1 2454 Intron XP_016857963.1
XM_017002475.1 2454 Intron XP_016857964.1
XM_017002476.1 2454 Intron XP_016857965.1
XM_017002477.1 2454 Intron XP_016857966.1
XM_017002478.1 2454 Intron XP_016857967.1
XM_017002479.1 2454 Intron XP_016857968.1

View Full Product Details