Product Details

SNP ID
rs147271626
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:153969369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCATTTATGGTGCCTGATTCC[A/T]GCATGGTCAGTGAGCTGCCCTTTGA
Phenotype
MIM: 607138 MIM: 604671 MIM: 604740
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CREB3L4 PubMed Links

Gene Details

Gene
CREB3L4
Gene Name
cAMP responsive element binding protein 3 like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001255978.1 695 Missense Mutation AGC,TGC S153C NP_001242907.1
NM_001255979.1 695 Missense Mutation AGC,TGC S153C NP_001242908.1
NM_001255980.1 695 Missense Mutation AGC,TGC S133C NP_001242909.1
NM_001255981.1 695 Missense Mutation AGC,TGC S133C NP_001242910.1
NM_130898.3 695 Missense Mutation AGC,TGC S153C NP_570968.1
XM_006711172.2 695 Missense Mutation AGC,TGC S133C XP_006711235.1
XM_017000372.1 695 Missense Mutation AGC,TGC S133C XP_016855861.1
Gene
JTB
Gene Name
jumping translocation breakpoint
There are no transcripts associated with this gene.

Gene
MIR6737
Gene Name
microRNA 6737
There are no transcripts associated with this gene.

Gene
SLC39A1
Gene Name
solute carrier family 39 member 1
There are no transcripts associated with this gene.

View Full Product Details