Product Details

SNP ID
rs147323508
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:1253979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAGGTTAAATACATACAATTGG[C/T]AAATTATAGAGCAATTCTGAATAAA
Phenotype
MIM: 616593
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM132A PubMed Links

Gene Details

Gene
FAM132A
Gene Name
family with sequence similarity 132 member A
There are no transcripts associated with this gene.

Gene
UBE2J2
Gene Name
ubiquitin conjugating enzyme E2 J2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058167.2 2066 UTR 3 NP_477515.2
NM_194315.1 2066 UTR 3 NP_919296.1
NM_194457.1 2066 UTR 3 NP_919439.1
NM_194458.1 2066 UTR 3 NP_919440.1
XM_005244718.3 2066 UTR 3 XP_005244775.1
XM_005244719.4 2066 UTR 3 XP_005244776.1
XM_006710333.3 2066 UTR 3 XP_006710396.1
XM_011540613.2 2066 UTR 3 XP_011538915.1
XM_011540614.2 2066 UTR 3 XP_011538916.1
XM_011540616.2 2066 UTR 3 XP_011538918.1
XM_011540617.2 2066 UTR 3 XP_011538919.1
XM_017000238.1 2066 UTR 3 XP_016855727.1
XM_017000239.1 2066 UTR 3 XP_016855728.1

View Full Product Details