Product Details

SNP ID
rs147413662
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:116392865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCTGTTCACAGCGGGCAGTTGC[A/C]GGAGATGCCTCTGAGTCAGCACTCT
Phenotype
MIM: 182310
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP1A1 PubMed Links

Gene Details

Gene
ATP1A1
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000701.7 1707 Silent Mutation GCA,GCC A448A NP_000692.2
NM_001160233.1 1707 Silent Mutation GCA,GCC A448A NP_001153705.1
NM_001160234.1 1707 Silent Mutation GCA,GCC A417A NP_001153706.1
XM_017001360.1 1707 Silent Mutation GCA,GCC A417A XP_016856849.1
XM_017001361.1 1707 Silent Mutation GCA,GCC A417A XP_016856850.1
Gene
ATP1A1-AS1
Gene Name
ATP1A1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details