Product Details

SNP ID
rs147739771
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:26057231 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAAAGTCAATGGCTCTCAGGGC[A/G]TCTGCTATGTGCTCTAAATCCAAAG
Phenotype
MIM: 606131
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM63 PubMed Links

Gene Details

Gene
TRIM63
Gene Name
tripartite motif containing 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032588.3 1091 Silent Mutation GAC,GAT D317D NP_115977.2
XM_017002559.1 1091 Silent Mutation GAC,GAT D317D XP_016858048.1

View Full Product Details