Product Details

SNP ID
rs147859973
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:6419333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGCCTTCTCTAGCTTCGAGCAG[C/T]TGGAGTTCTGCGCCCGGCCACGAGG
Phenotype
MIM: 606351 MIM: 609970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESPN PubMed Links

Gene Details

Gene
ESPN
Gene Name
espin
There are no transcripts associated with this gene.

Gene
HES2
Gene Name
hes family bHLH transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019089.4 248 Missense Mutation AAC,AGC N50S NP_061962.2

View Full Product Details