Product Details

SNP ID
rs148008982
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:156242277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCTATTGGCCCTGGCCGCACTT[C/T]GCATCGCTGGCCAGGCAGGTGAGTG
Phenotype
MIM: 112260 MIM: 614579 MIM: 609176 MIM: 610962
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BGLAP PubMed Links

Gene Details

Gene
BGLAP
Gene Name
bone gamma-carboxyglutamate protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199173.5 316 Missense Mutation CGC,TGC R16C NP_954642.1
Gene
PAQR6
Gene Name
progestin and adipoQ receptor family member 6
There are no transcripts associated with this gene.

Gene
PMF1
Gene Name
polyamine-modulated factor 1
There are no transcripts associated with this gene.

Gene
PMF1-BGLAP
Gene Name
PMF1-BGLAP readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199661.1 316 Intron NP_001186590.1
NM_001199662.1 316 Intron NP_001186591.1
NM_001199663.1 316 Intron NP_001186592.1
NM_001199664.1 316 Intron NP_001186593.1
Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details