Product Details

SNP ID
rs148014613
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:47438750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCAACTACTGGACGCTGGACCC[C/G]GAGTCGGCCGACATGTTCGACAACG
Phenotype
MIM: 602211
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FOXD2 PubMed Links

Gene Details

Gene
FOXD2
Gene Name
forkhead box D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004474.3 2734 Silent Mutation CCC,CCG P205P NP_004465.3
Gene
FOXD2-AS1
Gene Name
FOXD2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
LINC01389
Gene Name
long intergenic non-protein coding RNA 1389
There are no transcripts associated with this gene.

View Full Product Details