Product Details

SNP ID
rs148051047
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:28561378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTAAACTGAACTATGCCACTTA[C/T]GGGAAACAACCAGATAACAGGTAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRNAU1AP PubMed Links

Gene Details

Gene
TRNAU1AP
Gene Name
tRNA selenocysteine 1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017846.4 526 Silent Mutation TAC,TAT Y86Y NP_060316.1
XM_017001542.1 526 UTR 5 XP_016857031.1

View Full Product Details