Product Details

SNP ID
rs148126792
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:23362258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTCTGGTGTGTGATGAGATGA[C/G]AGCTCCGGCTGAAGCTTTTCCCACA
Phenotype
MIM: 611703
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF436 PubMed Links

Gene Details

Gene
ZNF436
Gene Name
zinc finger protein 436
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077195.1 1683 Missense Mutation TCT,TGT S375C NP_001070663.1
NM_030634.2 1683 Missense Mutation TCT,TGT S375C NP_085137.1
XM_011542201.1 1683 Missense Mutation TCT,TGT S357C XP_011540503.1
Gene
ZNF436-AS1
Gene Name
ZNF436 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details