Product Details

SNP ID
rs148149745
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:151770250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGAACTTCCAGAATGATCGGAA[A/C]GACAGAGGTGGGGATCTGCTGTTCT
Phenotype
MIM: 611824 MIM: 605138 MIM: 609501
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MRPL9 PubMed Links

Gene Details

Gene
MRPL9
Gene Name
mitochondrial ribosomal protein L9
There are no transcripts associated with this gene.

Gene
OAZ3
Gene Name
ornithine decarboxylase antizyme 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134939.1 2262 UTR 3 NP_001128411.1
NM_001301371.1 2262 UTR 3 NP_001288300.1
NM_016178.2 2262 UTR 3 NP_057262.2
Gene
TDRKH
Gene Name
tudor and KH domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083963.1 2262 Intron NP_001077432.1
NM_001083964.1 2262 Intron NP_001077433.1
NM_001083965.1 2262 Intron NP_001077434.1
NM_006862.3 2262 Intron NP_006853.2
XM_005244856.4 2262 Intron XP_005244913.1
XM_017000122.1 2262 Intron XP_016855611.1
XM_017000123.1 2262 UTR 3 XP_016855612.1
XM_017000124.1 2262 Intron XP_016855613.1
XM_017000125.1 2262 Intron XP_016855614.1
XM_017000126.1 2262 Intron XP_016855615.1
XM_017000127.1 2262 Intron XP_016855616.1
XM_017000128.1 2262 Intron XP_016855617.1
XM_017000129.1 2262 Intron XP_016855618.1
XM_017000130.1 2262 Intron XP_016855619.1

View Full Product Details