Product Details

SNP ID
rs148221696
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:202963144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGCAGTTCCTCTAAGTCCTCCC[G/T]CAAGATGATATCCTTTTCTGTCTGA
Phenotype
MIM: 607945 MIM: 608341
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADIPOR1 PubMed Links

Gene Details

Gene
ADIPOR1
Gene Name
adiponectin receptor 1
There are no transcripts associated with this gene.

Gene
CYB5R1
Gene Name
cytochrome b5 reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016243.2 738 Silent Mutation NP_057327.2

View Full Product Details