Product Details

SNP ID
rs148268889
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:247895686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCTCTTTGTGGTCATCCTGATC[A/G]CGTACCTCCTGACCCTCGTAGGCAA
Phenotype
MIM: 616729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2W3 PubMed Links

Gene Details

Gene
OR2W3
Gene Name
olfactory receptor family 2 subfamily W member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001957.2 100 Missense Mutation ACG,GCG T34A NP_001001957.2

View Full Product Details