Product Details

SNP ID
rs148579172
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:228425109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTAAAGTCCTGAGCGATCTCGC[G/T]CATCAGCCGCTGGAAGGGCAACTTG
Phenotype
MIM: 602820 MIM: 606123
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HIST3H3 PubMed Links

Gene Details

Gene
HIST3H3
Gene Name
histone cluster 3, H3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003493.2 217 Missense Mutation GGC,TGC G73C NP_003484.1
Gene
TRIM17
Gene Name
tripartite motif containing 17
There are no transcripts associated with this gene.

View Full Product Details