Product Details

SNP ID
rs148673499
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:203770038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTCTCTGCTCCTTCCTACCTAG[A/G]TGATCTCTGACATGTTACTTTACCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAX1 PubMed Links

Gene Details

Gene
LAX1
Gene Name
lymphocyte transmembrane adaptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136190.1 Intron NP_001129662.1
NM_001282878.1 Intron NP_001269807.1
NM_017773.3 Intron NP_060243.2
XM_006711397.3 Intron XP_006711460.1

View Full Product Details