Product Details

SNP ID
rs148676135
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:43273042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCCACCATGTGGCCCTGCTGC[A/G]CAGTGGTGGAGGGGCCGACGCCCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf210 PubMed Links

Gene Details

Gene
C1orf210
Gene Name
chromosome 1 open reading frame 210
There are no transcripts associated with this gene.

Gene
TMEM125
Gene Name
transmembrane protein 125
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320244.1 1052 Missense Mutation CAC,CGC H107R NP_001307173.1
NM_144626.2 1052 Missense Mutation CAC,CGC H107R NP_653227.1
XM_005270467.3 1052 Missense Mutation CAC,CGC H107R XP_005270524.1
XM_011540705.2 1052 Missense Mutation CAC,CGC H107R XP_011539007.1

View Full Product Details