Product Details
- SNP ID
-
rs148706291
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:89186420 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTCTTCTCCATTTGGGCCATGTA[C/T]TCCTGGAAGCTTCTCTCTTGAGCCT
- Phenotype
-
MIM: 612466
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GBP4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs655260] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GBP4
- Gene Name
- guanylate binding protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_052941.4 |
1736 |
Silent Mutation |
GAA,GAG |
E540E |
NP_443173.2 |
View Full Product Details