Product Details

SNP ID
rs148714772
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:24155849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCTACGGGTGAAAACTCAGCTA[C/T]GCTCATCTATATTCAGGTGGGTGTA
Phenotype
MIM: 607404
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFNLR1 PubMed Links

Gene Details

Gene
IFNLR1
Gene Name
interferon lambda receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170743.3 2784 UTR 3 NP_734464.1
NM_173064.2 2784 UTR 3 NP_775087.1
NM_173065.2 2784 UTR 3 NP_775088.1
XM_006710394.3 2784 UTR 3 XP_006710457.2
XM_017000479.1 2784 UTR 3 XP_016855968.1

View Full Product Details