Product Details

SNP ID
rs148938939
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:159062300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTATCTACTTTCAAATAATACTG[C/T]GCTGCTTCACATGTTGTACAGGATC
Phenotype
MIM: 604578 MIM: 147586
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIM2 PubMed Links

Gene Details

Gene
AIM2
Gene Name
absent in melanoma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004833.1 Intron NP_004824.1
XM_005245616.4 Intron XP_005245673.1
XM_017002848.1 Intron XP_016858337.1
Gene
IFI16
Gene Name
interferon gamma inducible protein 16
There are no transcripts associated with this gene.

View Full Product Details