Product Details

SNP ID
rs149011747
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:1293831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCGGCCGCGCTCACAGTGTCAC[A/G]ATGTCAGCGTTGGCCGCCTGCACTG
Phenotype
MIM: 601328
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAP3 PubMed Links

Gene Details

Gene
ACAP3
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030649.2 4899 Silent Mutation ATC,ATT I784I NP_085152.2
XM_005244715.2 4899 Silent Mutation ATC,ATT I751I XP_005244772.1
XM_005244717.3 4899 Silent Mutation ATC,ATT I443I XP_005244774.1
XM_011540606.2 4899 Silent Mutation ATC,ATT I794I XP_011538908.1
XM_011540607.1 4899 Silent Mutation ATC,ATT I794I XP_011538909.1
XM_011540608.2 4899 Silent Mutation ATC,ATT I762I XP_011538910.1
XM_011540609.2 4899 Silent Mutation ATC,ATT I761I XP_011538911.1
XM_017000233.1 4899 Silent Mutation ATC,ATT I595I XP_016855722.1
Gene
MIR6726
Gene Name
microRNA 6726
There are no transcripts associated with this gene.

Gene
SCNN1D
Gene Name
sodium channel epithelial 1 delta subunit
There are no transcripts associated with this gene.

View Full Product Details