Product Details

SNP ID
rs149381709
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:86863244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTGTTACAAAGCATTTTGTTAG[C/T]GGTATCAACAAATGCAGGAGGATGG
Phenotype
MIM: 606254
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEP15 PubMed Links

Gene Details

Gene
SEP15
Gene Name
15 kDa selenoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004261.3 1041 UTR 3 NP_004252.2
NM_203341.1 1041 UTR 3 NP_976086.1

View Full Product Details