Product Details

SNP ID
rs149398610
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:160291378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGGCCCGAGTTCTAGTAGGCGC[C/T]GAGCAAAGGTGGCAGCTGTCTTGAA
Phenotype
MIM: 601924 MIM: 600279
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPA PubMed Links

Gene Details

Gene
COPA
Gene Name
coatomer protein complex subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098398.1 3798 Missense Mutation CAG,CGG Q1135R NP_001091868.1
NM_004371.3 3798 Missense Mutation CAG,CGG Q1126R NP_004362.2
Gene
PEX19
Gene Name
peroxisomal biogenesis factor 19
There are no transcripts associated with this gene.

View Full Product Details