Product Details

SNP ID
rs149431183
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:158777333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCTCCCAAGGAGTGGAAGAAGTA[A/G]GTCTGAAGGAGGCATCCTGCAAAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6N1 PubMed Links

Gene Details

Gene
OR6N1
Gene Name
olfactory receptor family 6 subfamily N member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005185.1 303 Intron NP_001005185.1
XM_017000325.1 303 Intron XP_016855814.1
XM_017000326.1 303 Intron XP_016855815.1
XM_017000327.1 303 Intron XP_016855816.1
Gene
OR6N2
Gene Name
olfactory receptor family 6 subfamily N member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005278.1 303 Silent Mutation ACC,ACT T101T NP_001005278.1

View Full Product Details