Product Details

SNP ID
rs149555128
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:51790975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAAGAAACTCTTCTATCTTCTT[A/G]TCAACAACTTCAGAACTGAAACAAA
Phenotype
MIM: 602651 MIM: 606737
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NRDC PubMed Links

Gene Details

Gene
NRDC
Gene Name
nardilysin convertase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101662.1 3298 Silent Mutation GAC,GAT D992D NP_001095132.1
NM_001242361.1 3298 Silent Mutation GAC,GAT D928D NP_001229290.1
NM_002525.2 3298 Silent Mutation GAC,GAT D1060D NP_002516.2
XM_005270903.1 3298 Silent Mutation GAC,GAT D755D XP_005270960.1
XM_011541522.1 3298 Silent Mutation GAC,GAT D950D XP_011539824.1
XM_011541523.1 3298 Silent Mutation GAC,GAT D860D XP_011539825.1
XM_011541525.2 3298 Silent Mutation GAC,GAT D755D XP_011539827.1
XM_017001375.1 3298 Silent Mutation GAC,GAT D860D XP_016856864.1
Gene
OSBPL9
Gene Name
oxysterol binding protein like 9
There are no transcripts associated with this gene.

View Full Product Details