Product Details

SNP ID
rs149567639
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:23783782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGGAATATTTATAGGTGAAATG[A/C]TGTGTCTGGGATTGCTTTAAAATAT
Phenotype
MIM: 616143
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LYPLA2 PubMed Links

Gene Details

Gene
LYPLA2
Gene Name
lysophospholipase II
There are no transcripts associated with this gene.

Gene
PITHD1
Gene Name
PITH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020362.4 Intron NP_065095.2
XM_011541804.1 Intron XP_011540106.1
Gene
TCEB3-AS1
Gene Name
TCEB3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details