Product Details

SNP ID
rs149683559
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:101804416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATATGTGGAGGTTTTGGTGGAA[A/T]AGGAATAATACACCTTGGCTCCAGT
Phenotype
MIM: 607567
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OLFM3 PubMed Links

Gene Details

Gene
OLFM3
Gene Name
olfactomedin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288821.1 1521 Missense Mutation TAT,TTT Y420F NP_001275750.1
NM_001288823.1 1521 Missense Mutation TAT,TTT Y325F NP_001275752.1
NM_058170.3 1521 Missense Mutation TAT,TTT Y400F NP_477518.2
XM_017000240.1 1521 Missense Mutation TAT,TTT Y325F XP_016855729.1

View Full Product Details