Product Details

SNP ID
rs149957442
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:23362464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCTTCCCACACTCATCACACTT[A/G]TAGGGCCTCTCCCCTGTGTGGACTC
Phenotype
MIM: 611703
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF436 PubMed Links

Gene Details

Gene
ZNF436
Gene Name
zinc finger protein 436
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077195.1 1477 Silent Mutation TAC,TAT Y306Y NP_001070663.1
NM_030634.2 1477 Silent Mutation TAC,TAT Y306Y NP_085137.1
XM_011542201.1 1477 Silent Mutation TAC,TAT Y288Y XP_011540503.1
Gene
ZNF436-AS1
Gene Name
ZNF436 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details