Product Details

SNP ID
rs149971576
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:224227414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTCAGCAGTTTAAAAATTGTCC[C/T]TTTTCTTAAAGAGGAAGAAGGCATT
Phenotype
MIM: 602426
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927164 PubMed Links

Gene Details

Gene
LOC101927164
Gene Name
uncharacterized LOC101927164
There are no transcripts associated with this gene.

Gene
NVL
Gene Name
nuclear VCP-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243146.1 2845 UTR 3 NP_001230075.1
NM_001243147.1 2845 UTR 3 NP_001230076.1
NM_002533.3 2845 UTR 3 NP_002524.2
NM_206840.2 2845 UTR 3 NP_996671.1
XM_011544196.1 2845 UTR 3 XP_011542498.1
XM_011544198.1 2845 UTR 3 XP_011542500.1
XM_011544199.1 2845 UTR 3 XP_011542501.1
XM_011544200.1 2845 UTR 3 XP_011542502.1
XM_017001378.1 2845 UTR 3 XP_016856867.1
XM_017001379.1 2845 UTR 3 XP_016856868.1
XM_017001380.1 2845 Intron XP_016856869.1
XM_017001381.1 2845 UTR 3 XP_016856870.1
XM_017001382.1 2845 UTR 3 XP_016856871.1
XM_017001383.1 2845 UTR 3 XP_016856872.1
XM_017001384.1 2845 Intron XP_016856873.1
XM_017001385.1 2845 Intron XP_016856874.1
XM_017001386.1 2845 UTR 3 XP_016856875.1

View Full Product Details