Product Details

SNP ID
rs150169656
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:175004793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAAAACCAGCTGCTGTGGTTGC[C/T]CCCATTACAACGGGCTATACGGTGA
Phenotype
MIM: 606186 MIM: 611978 MIM: 609238
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACYBP PubMed Links

Gene Details

Gene
CACYBP
Gene Name
calcyclin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007214.1 567 Silent Mutation GCC,GCT A22A NP_001007215.1
NM_014412.2 567 Silent Mutation GCC,GCT A65A NP_055227.1
XM_017001046.1 567 Silent Mutation GCC,GCT A22A XP_016856535.1
Gene
LOC105371622
Gene Name
uncharacterized LOC105371622
There are no transcripts associated with this gene.

Gene
MRPS14
Gene Name
mitochondrial ribosomal protein S14
There are no transcripts associated with this gene.

Gene
RABGAP1L
Gene Name
RAB GTPase activating protein 1 like
There are no transcripts associated with this gene.

View Full Product Details