Product Details

SNP ID
rs150446205
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:8013730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATCTGGCTTTACTGGTCTAGGA[A/G]GTATGGGAACTCTGGGGGGAACCTC
Phenotype
MIM: 608069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERRFI1 PubMed Links

Gene Details

Gene
ERRFI1
Gene Name
ERBB receptor feedback inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018948.3 1132 Missense Mutation CCT,CTT P290L NP_061821.1
XM_005263477.2 1132 Missense Mutation CCT,CTT P239L XP_005263534.1
XM_006710697.2 1132 Missense Mutation CCT,CTT P290L XP_006710760.1
XM_011541596.2 1132 Missense Mutation CCT,CTT P215L XP_011539898.1

View Full Product Details