Product Details

SNP ID
rs150518910
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:36091810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCTCCCTGGCCTATAGCAGTGT[C/T]CAGGATGTGCAGAAGGTATTCAGGG
Phenotype
MIM: 610624 MIM: 120252 MIM: 608953
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADPRHL2 PubMed Links

Gene Details

Gene
ADPRHL2
Gene Name
ADP-ribosylhydrolase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017825.2 554 Silent Mutation GTC,GTT V167V NP_060295.1
XM_011541636.2 554 Silent Mutation GTC,GTT V13V XP_011539938.1
Gene
COL8A2
Gene Name
collagen type VIII alpha 2
There are no transcripts associated with this gene.

Gene
TEKT2
Gene Name
tektin 2
There are no transcripts associated with this gene.

View Full Product Details