Product Details

SNP ID
rs150604529
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:162497253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCCAAGAAAATAGATTGTAAAT[A/G]TTTCTTACCAGACCGAAGGTCTGTG
Phenotype
MIM: 608849
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UHMK1 PubMed Links

Gene Details

Gene
UHMK1
Gene Name
U2AF homology motif (UHM) kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184763.1 80 Missense Mutation ATA,ATG I1M NP_001171692.1
NM_144624.2 80 Intron NP_653225.2
NM_175866.4 80 Intron NP_787062.1

View Full Product Details