Product Details
- SNP ID
-
rs150610844
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:15626740 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCTTATGGCTTGAAAGATGGGGA[C/T]GTTGTGATTTTACGACAGAAGGAGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DDI2
PubMed Links
Gene Details
- Gene
- DDI2
- Gene Name
- DNA damage inducible 1 homolog 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032341.4 |
423 |
Silent Mutation |
GAC,GAT |
D70D |
NP_115717.3 |
View Full Product Details