Product Details
- SNP ID
-
rs150614243
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:155660882 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTCTAGCACCATCAGCCATGTGC[C/T]GCAGTTCTTCCTGGATGGATGGCAG
- Phenotype
-
MIM: 607860
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
YY1AP1
PubMed Links
Gene Details
- Gene
- YY1AP1
- Gene Name
- YY1 associated protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001198899.1 |
1634 |
Missense Mutation |
CAG,CGG |
Q332R |
NP_001185828.1 |
NM_001198900.1 |
1634 |
Missense Mutation |
CAG,CGG |
Q332R |
NP_001185829.1 |
NM_001198901.1 |
1634 |
Missense Mutation |
CAG,CGG |
Q343R |
NP_001185830.1 |
NM_001198902.1 |
1634 |
Missense Mutation |
CAG,CGG |
Q343R |
NP_001185831.1 |
NM_001198903.1 |
1634 |
Missense Mutation |
CAG,CGG |
Q481R |
NP_001185832.1 |
NM_001198904.1 |
1634 |
Missense Mutation |
CAG,CGG |
Q461R |
NP_001185833.1 |
NM_001198905.1 |
1634 |
Missense Mutation |
CAG,CGG |
Q323R |
NP_001185834.1 |
NM_001198906.1 |
1634 |
Missense Mutation |
AGC,GGC |
S416G |
NP_001185835.1 |
NM_018253.3 |
1634 |
Missense Mutation |
CAG,CGG |
Q332R |
NP_060723.2 |
NM_139118.2 |
1634 |
Missense Mutation |
CAG,CGG |
Q389R |
NP_620829.1 |
NM_139119.2 |
1634 |
Missense Mutation |
CAG,CGG |
Q343R |
NP_620830.1 |
NM_139121.2 |
1634 |
Missense Mutation |
CAG,CGG |
Q277R |
NP_620832.1 |
View Full Product Details