Product Details

SNP ID
rs150628229
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:178516640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAGGATCTAATGGACAAGGATT[G/T]TGATAAACTTCACGAATTTGTGGAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TEX35 PubMed Links

Gene Details

Gene
TEX35
Gene Name
testis expressed 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170722.1 354 Missense Mutation TGT,TTT C89F NP_001164193.1
NM_001170723.1 354 Missense Mutation TGT,TTT C81F NP_001164194.1
NM_001170724.1 354 Missense Mutation TGT,TTT C81F NP_001164195.1
NM_032126.4 354 Missense Mutation TGT,TTT C81F NP_115502.2

View Full Product Details