Product Details

SNP ID
rs150676565
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:38839562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGCTGGGATTCAGACTAGATGGC[A/G]TTTCGTGGCGTGCCATTGTGTGTCA
Phenotype
MIM: 608267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RRAGC PubMed Links

Gene Details

Gene
RRAGC
Gene Name
Ras related GTP binding C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271851.1 1266 Silent Mutation AAC,AAT N363N NP_001258780.1
NM_022157.3 1266 Silent Mutation AAC,AAT N397N NP_071440.1

View Full Product Details