Product Details

SNP ID
rs151266861
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:182056475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCAACTCAGAGGCAATGCGGAA[A/G]GCCCTGCCGCACTCGGCACAGAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF648 PubMed Links

Gene Details

Gene
ZNF648
Gene Name
zinc finger protein 648
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009992.1 5677 Silent Mutation NP_001009992.1
XM_017000301.1 5677 Silent Mutation XP_016855790.1
XM_017000302.1 5677 Silent Mutation XP_016855791.1

View Full Product Details