Product Details
- SNP ID
-
rs151305637
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:59132065 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGTCACATAACTTGTCCAAGGTT[A/G]CACAACTAGTTAACTGTTGGTGTTA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HSD52
PubMed Links
Gene Details
- Gene
- HSD52
- Gene Name
- uncharacterized LOC729467
There are no transcripts associated with this gene.
View Full Product Details