Product Details

SNP ID
rs151325808
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:159063597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTTACATTTCATTGTGTCCTC[A/G]TTTCTAACCCCCAGTACTTCCATTT
Phenotype
MIM: 604578 MIM: 147586
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AIM2 PubMed Links

Gene Details

Gene
AIM2
Gene Name
absent in melanoma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004833.1 1139 Silent Mutation NP_004824.1
XM_005245616.4 1139 Silent Mutation XP_005245673.1
XM_017002848.1 1139 Silent Mutation XP_016858337.1
Gene
IFI16
Gene Name
interferon gamma inducible protein 16
There are no transcripts associated with this gene.

View Full Product Details