Product Details

SNP ID
rs138502166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:388040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCACCCCTCTGGCTGCTCCTGC[A/G]GGTTCCCTGTCCAGGAAGAAGCGGT
Phenotype
MIM: 607898
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIB3 PubMed Links

Gene Details

Gene
TRIB3
Gene Name
tribbles pseudokinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301188.1 79 Silent Mutation GCA,GCG A10A NP_001288117.1
NM_001301190.1 79 Silent Mutation GCA,GCG A10A NP_001288119.1
NM_001301193.1 79 Silent Mutation GCA,GCG A10A NP_001288122.1
NM_001301196.1 79 Silent Mutation GCA,GCG A10A NP_001288125.1
NM_001301201.1 79 Silent Mutation GCA,GCG A37A NP_001288130.1
NM_021158.4 79 Silent Mutation GCA,GCG A10A NP_066981.2
XM_017027989.1 79 Silent Mutation GCA,GCG A37A XP_016883478.1

View Full Product Details