Product Details

SNP ID
rs138507928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:56474696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGCAGACTTTATAACAAAGATGC[C/T]GTCATTGAATTTCTCTTGGACAAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RTFDC1 PubMed Links

Gene Details

Gene
RTFDC1
Gene Name
replication termination factor 2 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283035.1 296 Silent Mutation GCC,GCT A91A NP_001269964.1
NM_001283036.1 296 Silent Mutation GCC,GCT A61A NP_001269965.1
NM_001283037.1 296 Silent Mutation GCC,GCT A61A NP_001269966.1
NM_016407.4 296 Silent Mutation GCC,GCT A61A NP_057491.2
XM_017027872.1 296 Silent Mutation GCC,GCT A42A XP_016883361.1

View Full Product Details