Product Details
- SNP ID
-
rs139386653
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:49984003 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGAGAGTCCCAGGGTGGCAGAG[C/G]TGACCTCCCTGTCAGATGAGGACAG
- Phenotype
-
MIM: 604238
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SNAI1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4647958] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNAI1
- Gene Name
- snail family transcriptional repressor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005985.3 |
346 |
Missense Mutation |
CTG,GTG |
L88V |
NP_005976.2 |
View Full Product Details