Product Details
- SNP ID
-
rs139400415
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:34727404 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTTTCAACTATCTCTTTGGGCTC[A/G]CTGCTGACAGAGGGAAGCAAAAAGT
- Phenotype
-
MIM: 605299
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NCOA6
PubMed Links
Gene Details
- Gene
- NCOA6
- Gene Name
- nuclear receptor coactivator 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001242539.2 |
6331 |
Silent Mutation |
AGC,AGT |
S1008S |
NP_001229468.1 |
NM_001318240.1 |
6331 |
Silent Mutation |
AGC,AGT |
S2001S |
NP_001305169.1 |
NM_014071.4 |
6331 |
Silent Mutation |
AGC,AGT |
S2001S |
NP_054790.2 |
XM_006723755.3 |
6331 |
Intron |
|
|
XP_006723818.1 |
XM_011528721.2 |
6331 |
Silent Mutation |
AGC,AGT |
S2006S |
XP_011527023.1 |
XM_011528726.2 |
6331 |
Silent Mutation |
AGC,AGT |
S1995S |
XP_011527028.1 |
XM_011528727.2 |
6331 |
Silent Mutation |
AGC,AGT |
S1994S |
XP_011527029.1 |
XM_017027739.1 |
6331 |
Silent Mutation |
AGC,AGT |
S2025S |
XP_016883228.1 |
XM_017027740.1 |
6331 |
Silent Mutation |
AGC,AGT |
S2025S |
XP_016883229.1 |
XM_017027741.1 |
6331 |
Silent Mutation |
AGC,AGT |
S2024S |
XP_016883230.1 |
XM_017027742.1 |
6331 |
Silent Mutation |
AGC,AGT |
S2020S |
XP_016883231.1 |
XM_017027743.1 |
6331 |
Intron |
|
|
XP_016883232.1 |
XM_017027744.1 |
6331 |
Silent Mutation |
AGC,AGT |
S1989S |
XP_016883233.1 |
XM_017027745.1 |
6331 |
Silent Mutation |
AGC,AGT |
S1977S |
XP_016883234.1 |
XM_017027746.1 |
6331 |
Intron |
|
|
XP_016883235.1 |
XM_017027747.1 |
6331 |
Silent Mutation |
AGC,AGT |
S1845S |
XP_016883236.1 |
XM_017027748.1 |
6331 |
Intron |
|
|
XP_016883237.1 |
XM_017027750.1 |
6331 |
Silent Mutation |
AGC,AGT |
S1032S |
XP_016883239.1 |
XM_017027751.1 |
6331 |
Silent Mutation |
AGC,AGT |
S1008S |
XP_016883240.1 |
View Full Product Details