Product Details

SNP ID
rs140046066
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35546695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCAGGAGGGAAGTGAGGACGTG[A/G]TAGTGTCAGCAGTGTCAAAGAATTG
Phenotype
MIM: 616971
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ERGIC3 PubMed Links

Gene Details

Gene
ERGIC3
Gene Name
ERGIC and golgi 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015966.2 Intron NP_057050.1
NM_198398.1 Intron NP_938408.1

View Full Product Details