Product Details

SNP ID
rs140145935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31605634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCTCAAGGAGCTGGTGCCCACC[C/T]TGCCCCAGAACCGCAAGGTGAGCAA
Phenotype
MIM: 600349
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ID1 PubMed Links

Gene Details

Gene
ID1
Gene Name
inhibitor of DNA binding 1, HLH protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002165.3 352 Silent Mutation CTG,TTG L83L NP_002156.2
NM_181353.2 352 Silent Mutation CTG,TTG L83L NP_851998.1
Gene
MIR3193
Gene Name
microRNA 3193
There are no transcripts associated with this gene.

View Full Product Details